Assay Details
Target Gene Details
Entrez Gene ID: | 100317 |
Gene Name: | expressed sequence AU040320 |
Gene Aliases: |
A730047D20Rik, Kiaa0319l |
Location: |
Chr.4:126753555-126869694 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AU040320 | NM_001035525.1 | 3 | 619 | NP_001030602.1 |
| NM_001035526.1 | 3 | 737 | NP_001030603.1 | |
| NM_133886.2 | 3 | 737 | NP_598647.1 | |
| AK031342.1 | 3 | 681 | ||
| AK043006.1 | 3 | 687 | BAC31432.1 | |
| AK049570.1 | 3 | 620 | BAC33818.1 | |
| AK084668.1 | 3 | 582 | BAC39244.1 | |
| AK170261.1 | 3 | 694 | BAE41669.1 | |
| AK177499.1 | 1 | 213 | ||
| BC028869.1 | 3 | 707 | AAH28869.1 | |
| BM942797.1 | 2 | 392 | ||
| CB249000.1 | 1 | 94 |

Genomic Map