Assay Details
Target Gene Details
Entrez Gene ID: | 242800 |
Gene Name: | tetratricopeptide repeat domain 34 |
Gene Aliases: |
4932418L02, B230396O12Rik |
Location: |
Chr.4:154833443-154867127 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc34 | NM_172878.3 | 5 | 1064 | NP_766466.3 |
| AK030055.1 | 5 | 1038 | BAC26761.1 |

Genomic Map