Assay Details
Target Gene Details
Entrez Gene ID: | 72421 |
Gene Name: | tetratricopeptide repeat domain 30B |
Gene Aliases: |
2510042P03Rik, Tcc30b |
Location: |
Chr.2:75935849-75938462 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc30b | NM_028235.1 | 1 | 2184 | NP_082511.1 |
| AK011097.1 | 1 | 2161 | BAB27394.1 | |
| AK147134.1 | 1 | 2134 | BAE27704.1 | |
| BC151060.1 | 1 | 2184 | ||
| BC151184.1 | 1 | 2184 | ||
| BC157929.1 | 1 | 2253 |

Genomic Map