Assay Details
Target Gene Details
Entrez Gene ID: | 54401 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta polypeptide |
Gene Aliases: |
1300003C17Rik |
Location: |
Chr.2:163995197-164018588 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ywhab | NM_018753.6 | 6 | 2132 | NP_061223.2 |
| AK002632.1 | 6 | 2151 | BAB22246.1 | |
| AK004872.1 | 6 | 2133 | BAB23631.1 | |
| AK083367.1 | 6 | 2122 | BAC38886.1 | |
| AK144061.1 | 6 | 2195 | BAE25678.1 | |
| AK158932.1 | 6 | 2035 | BAE34730.1 | |
| AK206846.1 | 1 | 193 | ||
| AK207996.1 | 1 | 193 | ||
| AK209116.1 | 1 | 193 | ||
| AK212148.1 | 1 | 193 | ||
| AK214843.1 | 1 | 193 | ||
| AK215126.1 | 1 | 193 | ||
| AK219468.1 | 1 | 193 |
More Information
Set Membership: |
|

Genomic Map