Assay Details
Target Gene Details
Entrez Gene ID: | 66641 |
Gene Name: | suppressor of IKBKE 1 |
Gene Aliases: |
2810005O12Rik, 5730470L24Rik, AI450236, AI839862, Sike, Sikeb |
Location: |
Chr.3:102995740-103003914 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sike1 | NM_025679.3 | 5 | 2056 | NP_079955.1 |
| AK033553.1 | 5 | 2060 | BAC28353.1 | |
| AK035590.1 | 5 | 2070 | BAC29116.1 |

Genomic Map