Assay Details
Target Gene Details
Entrez Gene ID: | 20674 |
Gene Name: | SRY (sex determining region Y)-box 2 |
Gene Aliases: |
Sox-2, lcc, ysb |
Location: |
Chr.3:34650005-34652461 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sox2 | NM_011443.3 | 1 | 1638 | NP_035573.3 |
| AK131933.1 | 1 | 1076 | BAE20882.1 | |
| U31967.1 | AAC31791.1 |
Target Gene Details
Entrez Gene ID: | 320478 |
Gene Name: | SOX2 overlapping transcript (non-protein coding) |
Gene Aliases: |
B230215L15Rik |
Location: |
Chr.3:34560381-34677993 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sox2ot | NR_015580.2 | |||
| AK045614.1 |
More Information
Set Membership: |
|

Genomic Map