Assay Details
Target Gene Details
Entrez Gene ID: | 12014 |
Gene Name: | BTB and CNC homology 2 |
Gene Aliases: |
E030004N02Rik |
Location: |
Chr.4:32238574-32586108 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bach2 | NM_001109661.1 | 3 | 934 | NP_001103131.1 |
| BC131642.1 | 3 | 842 | ||
| CX200493.1 | 1 | 487 |
Target Gene Details
Entrez Gene ID: | 414068 |
Gene Name: | BTB and CNC homology 2, opposite strand |
Gene Aliases: |
- |
Location: |
Chr.4:32559680-32571662 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bach2os | NR_026843.1 | |||
| AK144923.1 | BAE26138.1 |
More Information
Set Membership: |
|

Genomic Map