Assay Details
Target Gene Details
Entrez Gene ID: | 12014 |
Gene Name: | BTB and CNC homology 2 |
Gene Aliases: |
E030004N02Rik |
Location: |
Chr.4:32238574-32586108 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bach2 | NM_001109661.1 | 2 | 231 | NP_001103131.1 |
| AK162095.1 | 1 | 194 | BAE36721.1 | |
| BC099420.1 | 1 | 148 | AAH99420.1 | |
| BC131642.1 | 2 | 139 | ||
| CJ092609.1 | 3 | 588 |

Genomic Map