Assay Details
Target Gene Details
Entrez Gene ID: | 79235 |
Gene Name: | lecithin-retinol acyltransferase (phosphatidylcholine-retinol-O-acyltransferase) |
Gene Aliases: |
1300010A18Rik, AI449251 |
Location: |
Chr.3:82892582-82903974 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrat | NM_023624.4 | 1 | 405 | NP_076113.1 |
| AF255061.2 | 1 | 409 | AAF97787.1 | |
| AK004953.1 | 1 | 405 | BAB23696.1 | |
| BC141375.1 | 1 | 347 | ||
| BC141378.1 | 1 | 347 |
Target Gene Details
Entrez Gene ID: | 78212 |
Gene Name: | RNA binding motif protein 46, opposite strand |
Gene Aliases: |
4930444E23Rik, 4930564K09Rik, Lratos |
Location: |
Chr.3:82876705-82943638 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm46os | NR_040382.1 | |||
| AK019784.1 | ||||
| CJ047138.1 |
More Information
Set Membership: |
|

Genomic Map