Assay Details
Target Gene Details
Entrez Gene ID: | 12014 |
Gene Name: | BTB and CNC homology 2 |
Gene Aliases: |
E030004N02Rik |
Location: |
Chr.4:32238574-32586108 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bach2 | NM_001109661.1 | 5 | 7440 | NP_001103131.1 |

Genomic Map