Assay Details
Target Gene Details
Entrez Gene ID: | 230787 |
Gene Name: | thymocyte selection associated family member 2 |
Gene Aliases: |
ICB-1 |
Location: |
Chr.4:132781837-132796364 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Themis2 | NM_001033308.2 | 3 | 315 | NP_001028480.1 |
| AK137465.1 | 3 | 320 | ||
| AK149775.1 | 3 | 320 | ||
| AK150471.1 | 3 | 316 | BAE29588.1 | |
| AK153536.1 | 3 | 316 | BAE32073.1 | |
| AK155536.1 | 3 | 335 | BAE33313.1 | |
| AK171009.1 | 3 | 320 | BAE42180.1 | |
| AK171858.1 | 3 | 332 | ||
| BC013712.1 | 3 | 309 | AAH13712.1 | |
| BC151122.1 | 3 | 364 | ||
| BC151126.1 | 3 | 364 |

Genomic Map