Assay Details
Target Gene Details
Entrez Gene ID: | 230787 |
Gene Name: | thymocyte selection associated family member 2 |
Gene Aliases: |
ICB-1 |
Location: |
Chr.4:132781837-132796364 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Themis2 | NM_001033308.2 | 4 | 1556 | NP_001028480.1 |
| AK137465.1 | 4 | 1561 | ||
| AK149775.1 | 4 | 1561 | ||
| AK150471.1 | 4 | 1557 | BAE29588.1 | |
| AK153536.1 | 4 | 1557 | BAE32073.1 | |
| AK155536.1 | 4 | 1576 | BAE33313.1 | |
| AK171009.1 | 4 | 1561 | BAE42180.1 | |
| AK171858.1 | 4 | 1573 | ||
| BC013712.1 | 4 | 1550 | AAH13712.1 | |
| BC151122.1 | 4 | 1605 | ||
| BC151126.1 | 4 | 1605 |

Genomic Map