Assay Details
Target Gene Details
Entrez Gene ID: | 67758 |
Gene Name: | arylacetamide deacetylase (esterase) |
Gene Aliases: |
5033417E09Rik, AI265437, Aada |
Location: |
Chr.3:60031788-60040157 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aadac | NM_023383.1 | 5 | 840 | NP_075872.1 |
| AK008244.1 | 5 | 841 | ||
| BC019999.1 | 5 | 822 | AAH19999.1 | |
| BC054823.1 | 5 | 829 | AAH54823.1 |
More Information
Set Membership: |
|

Genomic Map