Assay Details
Target Gene Details
Entrez Gene ID: | 72162 |
Gene Name: | DEAH (Asp-Glu-Ala-His) box polypeptide 36 |
Gene Aliases: |
2810407E23Rik, AI452301, AU022184, Ddx36, mKIAA1488 |
Location: |
Chr.3:62468015-62507032 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhx36 | NM_028136.2 | 1 | 46 | NP_082412.2 |
| AF448804.1 | 1 | 46 | AAL47006.1 | |
| AK129373.1 | BAC98183.1 | |||
| BC006615.1 | 1 | 46 | ||
| BC138061.1 |
More Information
Set Membership: |
|

Genomic Map