Assay Details
Target Gene Details
Entrez Gene ID: | 94212 |
Gene Name: | phosphoprotein associated with glycosphingolipid microdomains 1 |
Gene Aliases: |
Cbp, F730007C19Rik, Pag |
Location: |
Chr.3:9687479-9833679 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pag1 | NM_001195031.1 | 9 | 3092 | NP_001181960.1 |
| NM_053182.5 | 8 | 3033 | NP_444412.2 | |
| AK089329.1 | 8 | 2985 | BAC40842.1 | |
| AK156000.1 | 9 | 3044 | BAE33539.1 |

Genomic Map