Assay Details
Target Gene Details
Entrez Gene ID: | 72274 |
Gene Name: | RIKEN cDNA 1700027H10 gene |
Gene Aliases: |
- |
Location: |
Chr.3:45416584-45439309 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700027H10Rik | NR_040594.1 | |||
| AK006417.1 |
Target Gene Details
Entrez Gene ID: | 18526 |
Gene Name: | protocadherin 10 |
Gene Aliases: |
6430521D13Rik, 6430703F07Rik, OL-pc, mKIAA1400 |
Location: |
Chr.3:45378398-45434579 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdh10 | NM_001098170.1 | 5 | 5008 | NP_001091640.1 |
| AK038615.1 | 5 | 3281 | ||
| AK046814.1 | 5 | 1910 |
More Information
Set Membership: |
|

Genomic Map