Assay Details
Target Gene Details
Entrez Gene ID: | 230235 |
Gene Name: | ferric-chelate reductase 1 like |
Gene Aliases: |
6430704M03Rik |
Location: |
Chr.4:56960136-56990391 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Frrs1l | NM_001142965.1 | 5 | 2847 | NP_001136437.1 |
| AK078331.1 | 4 | 2510 | BAC37221.1 |

Genomic Map