Assay Details
Target Gene Details
Entrez Gene ID: | 230393 |
Gene Name: | focadhesin |
Gene Aliases: |
Kiaa1797, mKIAA1797 |
Location: |
Chr.4:88094630-88411011 on Build GRCm38 |
Assay Gene Location: | Within Exon 44 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Focad | NM_001081184.1 | 44 | 5465 | NP_001074653.1 |
| AK122556.1 | 17 | 5659 | BAC65838.1 | |
| AK146138.1 | 15 | 1847 | BAE26929.1 | |
| AK146256.1 | 15 | 1847 | BAE27017.1 | |
| BC039622.1 | 6 | 846 | ||
| BC057079.1 | 33 | 3943 | AAH57079.1 |
Target Gene Details
Entrez Gene ID: | 66775 |
Gene Name: | 3-hydroxyacyl-CoA dehydratase 4 |
Gene Aliases: |
4933428I03Rik, Hcad4, Ptplad2 |
Location: |
Chr.4:88407687-88438935 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hacd4 | AK033780.1 | 1 | 1854 |

Genomic Map