Assay Details
Target Gene Details
Entrez Gene ID: | 18526 |
Gene Name: | protocadherin 10 |
Gene Aliases: |
6430521D13Rik, 6430703F07Rik, OL-pc, mKIAA1400 |
Location: |
Chr.3:45378398-45434579 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdh10 | NM_001098170.1 | 1 | 1164 | NP_001091640.1 |
| NM_001098171.1 | 1 | 1164 | NP_001091641.1 | |
| NM_001098172.1 | 1 | 1164 | NP_001091642.1 | |
| NM_011043.3 | 1 | 1164 | NP_035173.2 | |
| AF334801.1 | 1 | 400 | AAK57195.1 | |
| AK081621.1 | 1 | 1157 | ||
| AK081977.1 | 1 | 1157 | ||
| AK122503.1 | 1 | 1133 | BAC65785.1 | |
| BC065695.1 | 1 | 951 | AAH65695.1 | |
| U88549.1 | 1 | 401 | AAD00651.1 |
More Information
Set Membership: |
|

Genomic Map