Assay Details
Target Gene Details
Entrez Gene ID: | 56426 |
Gene Name: | programmed cell death 10 |
Gene Aliases: |
2410003B13Rik, Ccm3, Tfa15, Tfar15 |
Location: |
Chr.3:75516490-75556852 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdcd10 | NM_019745.4 | 8 | 1562 | NP_062719.2 |
| AK075926.1 | 8 | 1510 | BAC36058.1 | |
| AK077308.1 | 8 | 1539 | BAC36742.1 |

Genomic Map