Assay Details
Target Gene Details
Entrez Gene ID: | 320587 |
Gene Name: | transmembrane protein 88B |
Gene Aliases: |
A230069A22Rik |
Location: |
Chr.4:155781591-155785874 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem88b | NM_001033394.3 | 1 | 274 | NP_001028566.2 |
| AK035998.1 | 1 | 274 | ||
| AK158459.1 | 1 | 275 | BAE34518.1 | |
| BC147318.1 | 1 | 229 | ||
| BC147319.1 | 1 | 229 |

Genomic Map