Assay Details
Target Gene Details
Entrez Gene ID: | 332942 |
Gene Name: | predicted gene 853 |
Gene Aliases: |
OTTMUSG00000009560 |
Location: |
Chr.4:130209023-130222427 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm853 | NM_001034872.2 | 10 | 1812 | NP_001030044.1 |
| AK143920.1 | 10 | 1813 | BAE25605.1 |
More Information
Set Membership: |
|

Genomic Map