Assay Details
Target Gene Details
Entrez Gene ID: | 50798 |
Gene Name: | glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase |
Gene Aliases: |
2310066H07Rik, DMRV, GLCNE, IBM2, NM, Uae1 |
Location: |
Chr.4:44034075-44084177 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gne | NM_015828.3 | 12 | 3199 | NP_056643.3 |
| AK033507.1 | 3 | 1316 | BAC28328.1 | |
| AK033691.1 | 2 | 1118 | BAC28432.1 |
More Information
Set Membership: |
|

Genomic Map