Assay Details
Target Gene Details
Entrez Gene ID: | 53378 |
Gene Name: | syndecan binding protein |
Gene Aliases: |
MDA-9, Sycl, syntenin-1 |
Location: |
Chr.4:6365680-6396122 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdcbp | NM_001098227.1 | 9 | 2279 | NP_001091697.1 |
| NM_016807.2 | 9 | 2276 | NP_058087.2 | |
| AK028429.1 | 9 | 2252 | BAC25946.1 | |
| AK033412.1 | 9 | 2277 | BAC28275.1 | |
| AK171162.1 | 9 | 2251 | BAE42284.1 | |
| AK215213.1 | 1 | 124 | ||
| BB736053.1 | 1 | 139 | ||
| BY387130.1 | 1 | 78 |
More Information
Set Membership: |
|

Genomic Map