Assay Details
Target Gene Details
Entrez Gene ID: | 100504717 |
Gene Name: | predicted gene 12709 |
Gene Aliases: |
OTTMUSG00000008056 |
Location: |
Chr.4:102967266-102989755 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm12709 | NR_040444.1 | |||
| AK038336.1 |
Target Gene Details
Entrez Gene ID: | 73094 |
Gene Name: | SH3-domain GRB2-like (endophilin) interacting protein 1 |
Gene Aliases: |
3110007P09Rik, SGIP1alpha, gag |
Location: |
Chr.4:102759167-102972701 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgip1 | NM_001285852.1 | 26 | 3695 | NP_001272781.1 |
| NM_001285859.1 | 24 | 3110 | NP_001272788.1 | |
| NM_001285860.1 | 23 | 3050 | NP_001272789.1 | |
| NM_001285862.1 | 20 | 2957 | NP_001272791.1 | |
| NM_144906.2 | 24 | 3551 | NP_659155.1 | |
| AK032439.1 | 23 | 2841 | BAC27870.1 | |
| AK043018.1 | 24 | 2936 | BAC31435.1 | |
| AK148043.1 | 8 | 1638 | BAE28309.1 | |
| AK148302.1 | 20 | 2738 | BAE28466.1 | |
| BC017596.1 | 24 | 3323 | AAH17596.1 |
More Information
Set Membership: |
|

Genomic Map