Assay Details
Target Gene Details
Entrez Gene ID: | 94242 |
Gene Name: | tubulointerstitial nephritis antigen-like 1 |
Gene Aliases: |
1110021J17Rik, AZ-1, AZ1, Arg1, Lcn7, TARP, Tinagl |
Location: |
Chr.4:130165600-130175122 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tinagl1 | NM_001168333.1 | 12 | 1756 | NP_001161805.1 |
| NM_023476.3 | 12 | 1897 | NP_075965.2 | |
| AB047402.1 | 12 | 1898 | BAB20596.1 | |
| AB050626.1 | 12 | 1734 | BAC76038.1 | |
| AK003901.1 | 3 | 547 | ||
| AK219988.1 | 1 | 72 | ||
| BC005738.1 | 12 | 1724 | AAH05738.1 | |
| BC018539.1 | 12 | 1741 | AAH18539.1 |
More Information
Set Membership: |
|

Genomic Map