Assay Details
Target Gene Details
Entrez Gene ID: | 73750 |
Gene Name: | whirlin |
Gene Aliases: |
1110035G07Rik, AW122018, AW742671, C430046P22Rik, Dfnb31, wi |
Location: |
Chr.4:63414909-63496078 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Whrn | NM_001008791.2 | NP_001008791.1 | ||
| NM_001008792.2 | NP_001008792.1 | |||
| NM_001008793.2 | NP_001008793.1 | |||
| NM_028640.2 | NP_082916.1 | |||
| AK122523.1 | BAC65805.1 | |||
| AK157955.1 | BAE34281.1 | |||
| AY739114.1 | AAV87519.1 | |||
| AY739115.1 | AAV87520.1 | |||
| AY739116.1 | AAV87521.1 | |||
| AY739117.1 | AAV87522.1 | |||
| AY739118.1 | AAV87523.1 | |||
| AY739119.1 | AAV87524.1 | |||
| AY739120.1 | AAV87525.1 | |||
| AY739121.1 | 1 | 328 | AAV87526.1 | |
| AY739122.1 | 1 | 328 | AAV87527.1 | |
| BC150757.1 | ||||
| HQ148552.1 |
More Information
Set Membership: |
|

Genomic Map