Assay Details
Target Gene Details
Entrez Gene ID: | 12007 |
Gene Name: | alpha-2-glycoprotein 1, zinc |
Gene Aliases: |
Zag |
Location: |
Chr.5:137981521-137990233 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Azgp1 | NM_013478.2 | 2 | 269 | NP_038506.2 |
| AK005051.1 | 2 | 255 | BAB23777.1 | |
| AY248694.1 | 2 | 230 | AAO93093.1 | |
| BC061646.1 | 2 | 251 | AAH61646.1 | |
| BY101787.1 | 2 | 268 | ||
| BY705112.1 | 2 | 222 | ||
| D21059.1 | 2 | 230 | BAA04638.1 |
More Information
Set Membership: |
|

Genomic Map