Assay Details
Target Gene Details
Entrez Gene ID: | 73212 |
Gene Name: | RIKEN cDNA 3110082I17 gene |
Gene Aliases: |
AW411904 |
Location: |
Chr.5:139359739-139460534 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 3110082I17Rik | NM_028469.3 | NP_082745.2 | ||
| AK014271.1 | BAB29235.1 | |||
| AV470085.1 | ||||
| BU562762.1 |
Target Gene Details
Entrez Gene ID: | 80290 |
Gene Name: | G protein-coupled receptor 146 |
Gene Aliases: |
BC003323, PGR8 |
Location: |
Chr.5:139377698-139396415 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr146 | NM_001038703.2 | 2 | 1479 | NP_001033792.1 |
| NM_030258.4 | 2 | 1404 | NP_084534.2 | |
| AK046512.1 | 2 | 1427 | BAC32762.1 | |
| AK050034.1 | ||||
| AK087100.1 | BAC39803.1 | |||
| AK089232.1 | 2 | 1427 | BAC40804.1 | |
| AK137454.1 | 2 | 1481 | BAE23357.1 | |
| AK154478.1 | 2 | 1433 | BAE32614.1 | |
| AK154708.1 | 2 | 1425 | BAE32777.1 | |
| AK155212.1 | BAE33123.1 | |||
| AK155280.1 | 2 | 1491 | ||
| AK155876.1 | 2 | 1407 | BAE33478.1 | |
| AK172255.1 | 1 | 128 | ||
| AK218958.1 | 1 | 63 | ||
| BC003323.1 | 2 | 1375 | AAH03323.1 |
More Information
Set Membership: |
|

Genomic Map