Assay Details
Target Gene Details
Entrez Gene ID: | 329910 |
Gene Name: | acyl-CoA thioesterase 11 |
Gene Aliases: |
1110020M10Rik, 2010309H15Rik, AW060409, BFIT, BFIT1, Thea, Them1, mKIAA0707 |
Location: |
Chr.4:106744555-106807180 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acot11 | NM_025590.4 | 16 | 2321 | NP_079866.2 |
| AF416923.1 | 16 | 2259 | AAL40939.1 | |
| AK031588.1 | 16 | 2238 | BAC27464.1 | |
| AK038546.1 | 16 | 2259 | BAC30036.1 | |
| AK048670.1 | 16 | 2321 | BAC33416.1 | |
| AK052332.1 | 16 | 2337 | BAC34942.1 | |
| AK122350.1 | 15 | 2108 | BAC65632.1 | |
| AK170189.1 | 9 | 1344 | BAE41624.1 | |
| AK171255.1 | 9 | 1345 | BAE42347.1 | |
| BC042492.1 | 16 | 2256 | AAH42492.1 |
Target Gene Details
Entrez Gene ID: | 230579 |
Gene Name: | family with sequence simliarity 151, member A |
Gene Aliases: |
2010309H15Rik |
Location: |
Chr.4:106733915-106748292 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam151a | NM_146149.1 | 8 | 1513 | NP_666261.1 |
| AK008396.1 | 1 | 66 | ||
| AK008553.1 | 1 | 66 | ||
| AY168619.1 | 8 | 1513 | AAO38748.1 | |
| BC024426.1 | 8 | 1513 | AAH24426.1 | |
| BC024430.1 | 8 | 1513 | AAH24430.1 | |
| BC026682.1 | 8 | 1522 | AAH26682.1 |
More Information
Set Membership: |
|

Genomic Map