Assay Details
Target Gene Details
Entrez Gene ID: | 15207 |
Gene Name: | hairy and enhancer of split 3 (Drosophila) |
Gene Aliases: |
Hes-3, bHLHb43 |
Location: |
Chr.4:152285250-152291903 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hes3 | NM_008237.4 | 5 | 1611 | NP_032263.2 |
| AK090218.1 | 5 | 1612 | BAC41140.1 |
More Information
Set Membership: |
|

Genomic Map