Assay Details
Target Gene Details
Entrez Gene ID: | 231841 |
Gene Name: | BRCA1-associated ATM activator 1 |
Gene Aliases: |
AA881470, Baat1 |
Location: |
Chr.5:140704786-140719379 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brat1 | NM_001276287.1 | 5 | 864 | NP_001263216.1 |
| NM_181066.3 | 5 | 766 | NP_851411.1 | |
| NR_074085.1 | 5 | 766 | ||
| AK043212.1 | 5 | 729 | BAC31494.1 | |
| AK049321.1 | 5 | 755 | ||
| AK084987.1 | 5 | 755 | BAC39330.1 | |
| AK085091.1 | 5 | 755 | BAC39362.1 | |
| AK142387.1 | 4 | 1939 | ||
| AK154448.1 | 5 | 737 | BAE32593.1 | |
| AK170269.1 | 5 | 759 | BAE41674.1 | |
| AK188542.1 | ||||
| BC068301.1 | 5 | 830 | AAH68301.1 |

Genomic Map