Assay Details
Target Gene Details
Entrez Gene ID: | 329942 |
Gene Name: | CUB and Sushi multiple domains 2 |
Gene Aliases: |
B230311I21, B230378I17, Gm139, mKIAA1884 |
Location: |
Chr.4:127988016-128567666 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csmd2 | NM_001281955.1 | NP_001268884.1 | ||
| AK046385.1 | ||||
| AK140129.1 |
Target Gene Details
Entrez Gene ID: | 69317 |
Gene Name: | high-mobility group box 4 |
Gene Aliases: |
1700001F22Rik, AI326135 |
Location: |
Chr.4:128260212-128260895 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmgb4 | NM_027036.3 | 1 | 573 | NP_081312.2 |
| AK005588.1 | 1 | 572 | BAB24141.1 | |
| BC061030.1 | 1 | 569 | AAH61030.1 | |
| CA465179.1 | 1 | 572 | ||
| CN840046.1 |
More Information
Set Membership: |
|

Genomic Map