Assay Details
Target Gene Details
Entrez Gene ID: | 230119 |
Gene Name: | zinc finger and BTB domain containing 5 |
Gene Aliases: |
5930421I10, 9430083K24Rik, AI646847, mKIAA0354 |
Location: |
Chr.4:44991242-45012412 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb5 | NM_001163283.1 | 2 | 1270 | NP_001156755.1 |
| NM_001163284.1 | 2 | 2165 | NP_001156756.1 | |
| NM_173399.3 | 2 | 893 | NP_775575.2 | |
| AK031172.1 | 2 | 824 | ||
| AK129124.1 | 2 | 1728 | BAC97934.1 | |
| AK160003.1 | 2 | 1882 | BAE35551.1 | |
| AY635051.1 | 2 | 2165 | AAT47717.1 | |
| BC054551.1 | 2 | 751 | AAH54551.1 | |
| BC057545.1 | 2 | 2165 | AAH57545.1 |
More Information
Set Membership: |
|

Genomic Map