Assay Details
Target Gene Details
Entrez Gene ID: | 100041576 |
Gene Name: | predicted gene 3414 |
Gene Aliases: |
- |
Location: |
Chr.5:45719667-45727578 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm3414 | NR_027993.1 | 1 | 3955 | |
| AK141875.1 | 1 | 2999 | ||
| AK164990.1 | 1 | 1235 | BAE37993.1 |
Target Gene Details
Entrez Gene ID: | 209707 |
Gene Name: | ligand dependent nuclear receptor corepressor-like |
Gene Aliases: |
Mlr1 |
Location: |
Chr.5:45697184-45860115 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lcorl | NM_178142.5 | NP_835278.2 | ||
| AK042567.1 | BAC31295.1 | |||
| AK043498.1 | BAC31560.1 | |||
| AK043845.1 | BAC31678.1 | |||
| BC066151.1 | AAH66151.1 | |||
| BC138563.1 |
More Information
Set Membership: |
|

Genomic Map