Assay Details
Target Gene Details
Entrez Gene ID: | 80509 |
Gene Name: | mediator complex subunit 8 |
Gene Aliases: |
2210021A15Rik, AB041805, ARC32 |
Location: |
Chr.4:118409298-118415824 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med8 | NM_001290688.1 | 7 | 1626 | NP_001277617.1 |
| NM_020000.3 | 7 | 1666 | NP_064384.2 | |
| NM_173719.3 | 8 | 1564 | NP_776067.1 | |
| AK148025.1 | 7 | 1647 | BAE28295.1 | |
| BY429746.1 | 1 | 191 |
More Information
Set Membership: |
|

Genomic Map