Assay Details
Target Gene Details
Entrez Gene ID: | 85030 |
Gene Name: | tumor necrosis factor receptor superfamily, member 25 |
Gene Aliases: |
APO-3, DDR3, DR3, LARD, TR3, TRAMP, Tnfrsf12, WSL-1, WSL-LR, Wsl |
Location: |
Chr.4:152115528-152120119 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf25 | NM_001291010.1 | 10 | 1451 | NP_001277939.1 |
| NM_033042.4 | 7 | 1401 | NP_149031.2 | |
| AF329969.1 | 10 | 1455 | AAK11256.1 | |
| AK040632.1 | 7 | 2070 | ||
| BC017526.1 | 7 | 1401 | AAH17526.1 |

Genomic Map