Assay Details
Target Gene Details
Entrez Gene ID: | 66437 |
Gene Name: | fission 1 (mitochondrial outer membrane) homolog (yeast) |
Gene Aliases: |
2010003O14Rik, Ttc11 |
Location: |
Chr.5:136953264-136966234 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fis1 | NM_001163243.1 | 1 | 256 | NP_001156715.1 |
| CA322565.1 | 1 | 256 |

Genomic Map