Assay Details
Target Gene Details
Entrez Gene ID: | 74087 |
Gene Name: | solute carrier family 7, (cationic amino acid transporter, y+ system) member 13 |
Gene Aliases: |
0610009O04Rik, AGT-1, AGT1, XAT2 |
Location: |
Chr.4:19818727-19842213 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc7a13 | NM_028746.3 | 4 | 1398 | NP_083022.1 |
| AB072352.1 | 4 | 1381 | BAC00494.1 | |
| AJ417662.1 | 4 | 1347 | CAD10394.1 | |
| AK128901.1 | 4 | 1398 | ||
| AK143768.1 | 4 | 1400 | BAE25532.1 | |
| BC014684.1 | 4 | 1371 | AAH14684.1 |
More Information
Set Membership: |
|

Genomic Map