Assay Details
Target Gene Details
Entrez Gene ID: | 21949 |
Gene Name: | tumor necrosis factor (ligand) superfamily, member 8 |
Gene Aliases: |
CD153, CD30LG, Cd30l |
Location: |
Chr.4:63832820-63861681 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfsf8 | NM_009403.3 | 4 | 1670 | NP_033429.1 |
| AK030916.1 | 4 | 1671 | BAC27182.1 | |
| AK157270.1 | 4 | 1423 | BAE34022.1 | |
| L09754.1 | 4 | 1557 | AAA74595.1 |

Genomic Map