Assay Details
Target Gene Details
Entrez Gene ID: | 73212 |
Gene Name: | RIKEN cDNA 3110082I17 gene |
Gene Aliases: |
AW411904 |
Location: |
Chr.5:139359739-139460534 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 3110082I17Rik | NM_028469.3 | NP_082745.2 | ||
| AK014271.1 | BAB29235.1 | |||
| AV470085.1 | ||||
| BU562762.1 |
Target Gene Details
Entrez Gene ID: | 320197 |
Gene Name: | RIKEN cDNA D830046C22 gene |
Gene Aliases: |
- |
Location: |
Chr.5:139377697-139380053 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D830046C22Rik | NR_033147.1 | 1 | 1570 | |
| AK086028.1 | 1 | 1571 | BAC39596.1 | |
| AK087317.1 | 2 | 1163 | ||
| BC147358.1 | ||||
| BC147359.1 |
Target Gene Details
Entrez Gene ID: | 80290 |
Gene Name: | G protein-coupled receptor 146 |
Gene Aliases: |
BC003323, PGR8 |
Location: |
Chr.5:139377698-139396415 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr146 |
More Information
Set Membership: |
|

Genomic Map