Assay Details
Target Gene Details
Entrez Gene ID: | 22062 |
Gene Name: | transformation related protein 73 |
Gene Aliases: |
TAp73, Tp73, deltaNp73, p73 |
Location: |
Chr.4:154056249-154140208 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp73 | NM_001126330.1 | 12 | 3224 | NP_001119802.1 |
| NM_001126331.1 | 10 | 2936 | NP_001119803.1 | |
| NM_011642.3 | 14 | 3514 | NP_035772.2 | |
| AK014503.1 | 7 | 2203 | BAB29401.1 |
More Information
Set Membership: |
|

Genomic Map