Assay Details
Target Gene Details
Entrez Gene ID: | 110524 |
Gene Name: | diacylglycerol kinase, theta |
Gene Aliases: |
110kDa, DAGK, DAGK7, Dagk4, Dgkd |
Location: |
Chr.5:108647044-108660828 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgkq | NM_199011.1 | 23 | 4259 | NP_950176.1 |
| AK048764.1 | 22 | 4577 | ||
| AK132151.1 | 22 | 4165 | ||
| AK134745.1 | 10 | 3322 | BAE22264.1 | |
| AK143274.1 | 2 | 2266 | ||
| AK170840.1 | 20 | 4225 | ||
| BC062929.1 | 23 | 4259 | AAH62929.1 |
Target Gene Details
Entrez Gene ID: | 72392 |
Gene Name: | transmembrane protein 175 |
Gene Aliases: |
3010001K23Rik, AI504381 |
Location: |
Chr.5:108629801-108647770 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem175 | NM_001163531.1 | 11 | 2523 | NP_001157003.1 |
| NM_001163532.1 | 11 | 2526 | NP_001157004.1 | |
| NM_028223.3 | 11 | 2633 | NP_082499.3 | |
| AK034579.1 | 11 | 2502 | BAC28761.1 |
More Information
Set Membership: |
|

Genomic Map