Assay Details
Target Gene Details
Entrez Gene ID: | 12578 |
Gene Name: | cyclin-dependent kinase inhibitor 2A |
Gene Aliases: |
ARF-INK4a, Arf, INK4a-ARF, Ink4a/Arf, MTS1, Pctr1, p16, p16(INK4a), p16INK4a, p19<ARF>, p19ARF |
Location: |
Chr.4:89274473-89294619 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdkn2a | NM_001040654.1 | 2 | 491 | NP_001035744.1 |
| NM_009877.2 | 2 | 570 | NP_034007.1 | |
| AB221638.1 | 2 | 475 | ||
| AF044335.1 | 2 | 491 | AAC08962.1 | |
| AF044336.1 | 2 | 491 | AAC08963.1 | |
| AK155307.1 | 2 | 480 | BAE33180.1 | |
| BC058190.1 | 2 | 490 | ||
| EU071702.1 | 2 | 342 | ||
| L76092.1 | 2 | 516 | AAC42080.1 | |
| L76150.1 | 2 | 488 | AAA85453.1 | |
| S80650.1 | 2 | 516 | AAB35770.1 |

Genomic Map