Assay Details
Target Gene Details
Entrez Gene ID: | 320790 |
Gene Name: | chromodomain helicase DNA binding protein 7 |
Gene Aliases: |
A730019I05Rik, Cycn, Cyn, Dz, Edy, Flo, GENA 47, GENA 60, Gena 52, Lda, Mt, Obt, Todo, WBE1, Whi, metis |
Location: |
Chr.4:8690406-8868449 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chd7 | NM_001277149.1 | 2 | 675 | NP_001264078.1 |
| AK035953.1 | 2 | 525 | ||
| AK042727.1 | 2 | 376 | BAC31343.1 | |
| AK171051.1 | 2 | 453 | BAE42213.1 | |
| AK171857.1 | 3 | 462 | BAE42701.1 | |
| AK172025.1 | 2 | 677 | BAE42783.1 |

Genomic Map