Assay Details
Target Gene Details
Entrez Gene ID: | 13486 |
Gene Name: | down-regulator of transcription 1 |
Gene Aliases: |
1700121L09Rik, Dr1l, NC2, NC2beta |
Location: |
Chr.5:108268897-108280521 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dr1 | NM_026106.4 | 1 | 462 | NP_080382.2 |
AK007230.1 | 1 | 442 | ||
AK018040.1 | 1 | 385 | ||
AK031057.1 | 1 | 396 | BAE20474.1 | |
AK075713.1 | 1 | 462 | BAC35903.1 | |
AK090113.1 | 1 | 444 | BAC41099.1 | |
AK139866.1 | 1 | 392 | BAE24166.1 | |
AK144749.1 | 1 | 456 | BAE26046.1 | |
AK146573.1 | 1 | 425 | BAE27269.1 | |
AK219205.1 | 1 | 115 | ||
BC013461.1 | AAH13461.1 |
