Assay Details
Target Gene Details
Entrez Gene ID: | 12733 |
Gene Name: | chloride channel Ka |
Gene Aliases: |
C75963, CLC-K1, Clcnk1 |
Location: |
Chr.4:141384611-141398706 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clcnka | NM_001146307.1 | 19 | 2235 | NP_001139779.1 |
| NM_024412.3 | 20 | 2239 | NP_077723.3 | |
| BB743618.1 | 2 | 269 | ||
| BC037077.1 | 20 | 2243 | AAH37077.1 | |
| BG061970.1 | 1 | 171 |
More Information
Set Membership: |
|

Genomic Map