Assay Details
Target Gene Details
Entrez Gene ID: | 18781 |
Gene Name: | phospholipase A2, group IIC |
Gene Aliases: |
- |
Location: |
Chr.4:138725325-138744575 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g2c | NM_008868.3 | 6 | 803 | NP_032894.2 |
| BC029347.1 | 5 | 648 | AAH29347.1 | |
| U18119.1 | 5 | 629 | AAC52936.1 |
More Information
Set Membership: |
|

Genomic Map