Assay Details
Target Gene Details
Entrez Gene ID: | 622665 |
Gene Name: | coiled-coil domain containing 17 |
Gene Aliases: |
- |
Location: |
Chr.4:116596730-116600266 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc17 | NM_001037916.3 | 13 | 1692 | NP_001033005.1 |
| AK003161.1 | 3 | 283 | BAC25021.1 | |
| AK029257.1 | 13 | 1694 | BAC26359.1 | |
| AK043689.1 | 6 | 901 | ||
| AK076542.1 | 12 | 1663 | ||
| BC150874.1 | 11 | 1479 | ||
| BC172023.1 | 11 | 1479 |

Genomic Map