Assay Details
Target Gene Details
Entrez Gene ID: | 21349 |
Gene Name: | T cell acute lymphocytic leukemia 1 |
Gene Aliases: |
SCL/tal-1, Scl, bHLHa17, tal-1 |
Location: |
Chr.4:115056426-115071755 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tal1 | NM_001287388.1 | 3 | 425 | NP_001274317.1 |
| NM_011527.3 | 3 | 408 | NP_035657.1 | |
| AK144164.1 | 2 | 378 | ||
| AK157852.1 | 3 | 426 | BAE34231.1 | |
| BC063060.1 | 3 | 408 | AAH63060.1 | |
| M59764.1 | 2 | 356 | AAA40097.1 |
More Information
Set Membership: |
|

Genomic Map